2,3-Dibromo-1-chloro-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

177683-1G 177683-500MG 177683-5G
77465-978EA 746.75 USD
77465-978 77478-722 77488-942
2,3-Dibromo-1-chloro-4-fluorobenzene ≥95%
2,3-Dibromo-1-chloro-4-fluorobenzene

Precaución: For research use only

Formula: C₆H₂Br₂ClF
Storage Temperature: Ambient
MDL Number: MFCD28750472
CAS Number: 1805525-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR