2-Bromo-1,4-diethylbenzene ≥95%

Supplier: AOBChem USA

Warning

177513-1G 177513-500MG 177513-5G 177513-25G
77465-974EA 287.37 USD
77465-974 77478-720 77488-938 77499-428
2-Bromo-1,4-diethylbenzene ≥95%
2-Bromo-1,4-diethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃Br
MW: 213.1142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394811
CAS Number: 52076-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR