2-Bromo-1,3,5-triethylbenzene ≥96%

Supplier: AOBChem USA

Warning

177504-1G 177504-500MG 177504-5G 177504-25G
77465-970EA 203.94 USD
77465-970 77478-716 77488-934 77499-424
2-Bromo-1,3,5-triethylbenzene ≥96%
2-Bromo-1,3,5-triethylbenzene

Precaución: For research use only

Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045018
CAS Number: 91-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR