2-Fluoro-3-(trifluoromethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17708-1G 17708-500MG 17708-5G 17708-25G
77465-966EA 75.19 USD
77465-966 77478-712 77488-930 77499-420
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid ≥95%
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid

Precaución: For research use only

Formula: C₇H₅BF₄O₃
Storage Temperature: Ambient
MDL Number: MFCD11045053
CAS Number: 881402-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR