1-Bromo-3-(methoxymethoxy)-5-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

17700-1G 17700-500MG 17700-5G 17700-25G
77465-962EA 284.28 USD
77465-962 77478-708 77488-926 77499-416
1-Bromo-3-(methoxymethoxy)-5-(trifluoromethoxy)benzene ≥97%
1-Bromo-3-(methoxymethoxy)-5-(trifluoromethoxy)benzene

Precaución: For research use only

Formula: C₉H₈BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD32879356
CAS Number: 2158298-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR