9-(4-Methoxyphenyl)-9H-fluoren-9-ol ≥95%

Supplier: AOBChem USA

Warning

176914-1G 176914-500MG 176914-5G 176914-25G
77465-958EA 415.09 USD
77465-958 77478-704 77488-922 77499-414
9-(4-Methoxyphenyl)-9H-fluoren-9-ol ≥95%
9-(4-Methoxyphenyl)-9H-fluoren-9-ol

Precaución: For research use only

Formula: C₂₀H₁₆O₂
MW: 288.3398 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30561925
CAS Number: 57028-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR