3'-bromo-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥96%

Supplier: AOBChem USA

Warning

176898-1G 176898-500MG 176898-5G 176898-25G
77465-954EA 382.13 USD
77465-954 77478-700 77488-918 77499-410
3'-bromo-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥96%
3'-bromo-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃BrO
MW: 253.1350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34855057
CAS Number: 2005737-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR