2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

17638-1G 17638-500MG 17638-5G 17638-25G
77465-946EA 380.07 USD
77465-946 77478-692 77488-910 77499-402
2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester ≥95%
2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₃H₂₁BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12923387
CAS Number: 1111637-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR