3-Bromo-5-chloro-benzene-1,2-diol ≥97%

Supplier: AOBChem USA

Warning

17599-1G 17599-500MG 17599-5G 17599-25G
77465-942EA 408.91 USD
77465-942 77478-690 77488-906 77499-398
3-Bromo-5-chloro-benzene-1,2-diol ≥97%
3-Bromo-5-chloro-benzene-1,2-diol

Precaución: For research use only

Formula: C₆H₄BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD23701700
CAS Number: 180995-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR