5-Methylthiophene-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Methyl-3-thiophenecarboxaldehyde

175805-1G 175805-500MG 175805-5G 175805-25G
77465-938EA 111.24 USD
77465-938 77478-686 77488-902 77499-394
5-Methylthiophene-3-carbaldehyde ≥95%
5-Methylthiophene-3-carbaldehyde

Precaución: For research use only

Formula: C₆H₆OS
Storage Temperature: Ambient
MDL Number: MFCD07792887
CAS Number: 29421-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR