2-Chloro-α,α-dimethyl-3-pyridinemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
175394-1G
175394-500MG
175394-5G
77465-930EA
670.53
USD
77465-930
77478-678
77488-892
2-Chloro-α,α-dimethyl-3-pyridinemethanol ≥95%
2-Chloro-α,α-dimethyl-3-pyridinemethanol
Precaución: For research use only
Formula:
C₈H₁₀ClNO Storage Temperature: Ambient |
MDL Number:
MFCD27948451 CAS Number: 267003-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...