1-Bromo-3-(4-nitrophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

175232-1G 175232-500MG 175232-5G 175232-25G
77465-926EA 328.57 USD
77465-926 77478-674 77488-888 77499-386
1-Bromo-3-(4-nitrophenoxy)benzene ≥97%
1-Bromo-3-(4-nitrophenoxy)benzene

Precaución: For research use only

Formula: C₁₂H₈BrNO₃
MW: 294.1008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05991358
CAS Number: 2303-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR