1-(3-Fluoro-5-methoxyphenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

175227-1G 175227-500MG 175227-5G 175227-25G
77465-922EA 397.58 USD
77465-922 77478-670 77488-884 77499-382
1-(3-Fluoro-5-methoxyphenyl)-2-methylbutan-1-ol ≥95%
1-(3-Fluoro-5-methoxyphenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
MW: 212.2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854570
CAS Number: 1994548-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR