1-(4-Fluorophenyl)-2-methylbutan-1-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
175223-1G
175223-500MG
175223-5G
175223-25G
77465-918EA
415.09
USD
77465-918
77478-666
77488-880
77499-378
1-(4-Fluorophenyl)-2-methylbutan-1-ol ≥97%
1-(4-Fluorophenyl)-2-methylbutan-1-ol
Precaución: For research use only
Formula:
C₁₁H₁₅FO MW: 182.2346 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12786841 CAS Number: 774-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...