1-(4-Methoxyphenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

175219-1G 175219-500MG 175219-5G 175219-25G
77465-914EA 297.67 USD
77465-914 77478-662 77488-876 77499-374
1-(4-Methoxyphenyl)-2-methylbutan-1-ol ≥95%
1-(4-Methoxyphenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
MW: 194.2701 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786933
CAS Number: 14248-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR