1-(3-Bromo-2-fluorophenyl)-2-methylbutan-1-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
175213-1G
175213-500MG
175213-5G
175213-25G
77465-910EA
397.58
USD
77465-910
77478-658
77488-872
77499-370
1-(3-Bromo-2-fluorophenyl)-2-methylbutan-1-ol ≥97%
1-(3-Bromo-2-fluorophenyl)-2-methylbutan-1-ol
Precaución: For research use only
Formula:
C₁₁H₁₄BrFO MW: 261.1306 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29938738 CAS Number: 1693655-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...