4-(2-Fluoro-5-methylphenyl)tetrahydro-2H-thiopyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

175205-1G 175205-500MG 175205-5G
77465-902EA 447.02 USD
77465-902 77478-650 77488-864
4-(2-Fluoro-5-methylphenyl)tetrahydro-2H-thiopyran-4-ol ≥97%
4-(2-Fluoro-5-methylphenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₅FOS
MW: 226.3103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854564
CAS Number: 2006461-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR