1-(5-Bromo-2-fluorophenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175190-1G 175190-500MG 175190-5G 175190-25G
77465-894EA 415.09 USD
77465-894 77478-642 77488-856 77499-360
1-(5-Bromo-2-fluorophenyl)-3-methylbutan-1-ol ≥97%
1-(5-Bromo-2-fluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.1306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19601823
CAS Number: 1344295-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR