4-(2,6-Difluorophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

175167-1G 175167-500MG 175167-5G 175167-25G
77465-890EA 312.09 USD
77465-890 77478-638 77488-852 77499-356
4-(2,6-Difluorophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%
4-(2,6-Difluorophenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₂F₂OS
MW: 230.2741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854554
CAS Number: 939801-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR