2,4,6-Trifluoro-1-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

17503-1G 17503-500MG 17503-5G 17503-25G
77465-886EA 219.39 USD
77465-886 77478-634 77488-848 77499-352
2,4,6-Trifluoro-1-(methoxymethoxy)benzene ≥97%
2,4,6-Trifluoro-1-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇F₃O₂
MW: 192.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976934
CAS Number: 2179038-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR