4-(4-Fluorophenyl)tetrahydro-2H-thiopyran-4-ol ≥96%

Supplier: AOBChem USA

Warning

174761-1G 174761-500MG 174761-5G
77465-882EA 483.07 USD
77465-882 77478-630 77488-844
4-(4-Fluorophenyl)tetrahydro-2H-thiopyran-4-ol ≥96%
4-(4-Fluorophenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₃FOS
MW: 212.2837 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30477406
CAS Number: 939801-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR