2',4',6'-trifluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

174755-1G 174755-500MG 174755-5G
77465-878EA 546.93 USD
77465-878 77478-626 77488-840
2',4',6'-trifluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
2',4',6'-trifluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: For research use only

Formula: C₁₃H₇F₃O
MW: 236.1892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12860947
CAS Number: 1182893-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR