3-Benzyloxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17442-1G 17442-500MG 17442-5G 17442-25G
77465-874EA 333.72 USD
77465-874 77478-622 77488-836 77499-342
3-Benzyloxy-2-fluorobenzaldehyde ≥97%
3-Benzyloxy-2-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₄H₁₁FO₂
MW: 230.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110192
CAS Number: 103438-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR