5-Bromo-2-ethoxy-3-iodo-pyridine ≥97%

Supplier: AOBChem USA

Warning

17418-1G 17418-500MG 17418-5G 17418-25G
77465-870EA 100.94 USD
77465-870 77478-620 77488-832 77499-338
5-Bromo-2-ethoxy-3-iodo-pyridine ≥97%
5-Bromo-2-ethoxy-3-iodo-pyridine

Precaución: For research use only

Formula: C₇H₇BrINO
Storage Temperature: Ambient
MDL Number: MFCD07366718
CAS Number: 848243-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR