1-Ethoxy-4-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

173219-1G 173219-500MG 173219-5G 173219-25G
77465-862EA 397.58 USD
77465-862 77478-614 77488-824 77499-328
1-Ethoxy-4-[(1-methylethyl)thio]benzene ≥95%
1-Ethoxy-4-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621832
CAS Number: 82961-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR