1-Methoxy-4-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

173185-1G 173185-500MG 173185-5G 173185-25G
77465-858EA 127.72 USD
77465-858 77478-610 77488-820 77499-326
1-Methoxy-4-[(1-methylethyl)thio]benzene ≥95%
1-Methoxy-4-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056033
CAS Number: 7205-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR