4-(Ethylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

173179-1G 173179-500MG 173179-5G 173179-25G
77465-854EA 245.14 USD
77465-854 77478-606 77488-816 77499-324
4-(Ethylthio)benzonitrile ≥97%
4-(Ethylthio)benzonitrile

Precaución: For research use only

Formula: C₉H₉NS
Storage Temperature: Ambient
MDL Number: MFCD11136354
CAS Number: 86047-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR