5-Chloro-2-(propylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

173001-1G 173001-500MG 173001-5G 173001-25G
77465-850EA 397.58 USD
77465-850 77478-604 77488-808 77499-316
5-Chloro-2-(propylthio)pyridine ≥97%
5-Chloro-2-(propylthio)pyridine

Precaución: For research use only

Formula: C₈H₁₀ClNS
MW: 187.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21641937
CAS Number: 1466129-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR