6-Bromo-4-fluoro-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

172857-1G 172857-500MG 172857-5G 172857-25G
77465-846EA 215.27 USD
77465-846 77478-600 77488-804 77499-312
6-Bromo-4-fluoro-2-methyl-2H-indazole ≥97%
6-Bromo-4-fluoro-2-methyl-2H-indazole

Precaución: For research use only

Formula: C₈H₆BrFN₂
MW: 229.0490 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31743201
CAS Number: 2092036-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR