2',6'-Dichloro-3,4'-bipyridine ≥95%

Supplier: AOBChem USA

Warning

171481-1G 171481-500MG 171481-5G
77465-842EA 575.77 USD
77465-842 77478-596 77488-798
2',6'-Dichloro-3,4'-bipyridine ≥95%
2',6'-Dichloro-3,4'-bipyridine

Precaución: For research use only

Formula: C₁₀H₆Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD08063170
CAS Number: 867373-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR