7-Methoxybenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

171196-1G 171196-500MG 171196-5G 171196-25G
77465-838EA 163.77 USD
77465-838 77478-592 77488-794 77499-302
7-Methoxybenzo[b]thiophene ≥95%
7-Methoxybenzo[b]thiophene

Precaución: For research use only

Formula: C₉H₈OS
MW: 164.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706230
CAS Number: 88791-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR