4-Chloro-2-(1,1-dimethylethyl)-5-methylphenol ≥95%

Supplier: AOBChem USA

Warning

170590-1G 170590-500MG 170590-5G 170590-25G
77465-834EA 149.35 USD
77465-834 77478-588 77488-788 77499-296
4-Chloro-2-(1,1-dimethylethyl)-5-methylphenol ≥95%
4-Chloro-2-(1,1-dimethylethyl)-5-methylphenol

Precaución: For research use only

Formula: C₁₁H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD01731707
CAS Number: 30894-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR