3-(1,1-Dimethylethyl)-2-ethoxy-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

170551-1G 170551-500MG 170551-5G 170551-25G
77465-830EA 415.09 USD
77465-830 77478-584 77488-784 77499-294
3-(1,1-Dimethylethyl)-2-ethoxy-5-methylbenzoic acid ≥95%
3-(1,1-Dimethylethyl)-2-ethoxy-5-methylbenzoic acid

Precaución: For research use only

Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28580496
CAS Number: 1367822-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR