1-(3-Chloro-2-fluorophenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170119-1G 170119-500MG 170119-5G 170119-25G
77465-826EA 397.58 USD
77465-826 77478-580 77488-778 77499-290
1-(3-Chloro-2-fluorophenyl)-2-methylbutan-1-ol ≥95%
1-(3-Chloro-2-fluorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.6796 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21929203
CAS Number: 1536756-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR