1-(5-Bromo-2-fluorophenyl)butan-1-ol ≥97%

Supplier: AOBChem USA

Warning

170109-1G 170109-500MG 170109-5G 170109-25G
77465-822EA 415.09 USD
77465-822 77478-576 77488-774 77499-286
1-(5-Bromo-2-fluorophenyl)butan-1-ol ≥97%
1-(5-Bromo-2-fluorophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.1040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19601821
CAS Number: 1197943-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR