1-(2-Chlorophenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170098-1G 170098-500MG 170098-5G 170098-25G
77465-818EA 169.95 USD
77465-818 77478-572 77488-770 77499-284
1-(2-Chlorophenyl)-2-methylbutan-1-ol ≥95%
1-(2-Chlorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅ClO
MW: 198.6892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18871695
CAS Number: 124285-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR