1-(4-Bromophenyl)-2-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

170088-1G 170088-500MG 170088-5G 170088-25G
77465-814EA 297.67 USD
77465-814 77478-568 77488-766 77499-280
1-(4-Bromophenyl)-2-methylbutan-1-ol ≥97%
1-(4-Bromophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.1402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786806
CAS Number: 261783-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR