1-(2,4-Difluorophenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170084-1G 170084-500MG 170084-5G 170084-25G
77465-810EA 297.67 USD
77465-810 77478-564 77488-762 77499-276
1-(2,4-Difluorophenyl)-3-methylbutan-1-ol ≥95%
1-(2,4-Difluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄F₂O
MW: 200.2250 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787301
CAS Number: 1249525-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR