1-(3-Bromophenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170080-1G 170080-500MG 170080-5G 170080-25G
77465-806EA 415.09 USD
77465-806 77478-560 77488-758 77499-272
1-(3-Bromophenyl)-3-methylbutan-1-ol ≥95%
1-(3-Bromophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.1402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153606
CAS Number: 1183885-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR