1-(2-Fluorophenyl)-3-methylbutan-1-ol ≥96%

Supplier: AOBChem USA

Warning

170074-1G 170074-500MG 170074-5G 170074-25G
77465-802EA 415.09 USD
77465-802 77478-556 77488-754 77499-268
1-(2-Fluorophenyl)-3-methylbutan-1-ol ≥96%
1-(2-Fluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.2346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16302230
CAS Number: 765306-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR