3-Methyl-1-phenylbutan-1-ol ≥96%

Supplier: AOBChem USA

Warning

170070-1G 170070-500MG 170070-5G 170070-25G
77465-798EA 267.8 USD
77465-798 77478-552 77488-750 77499-264
3-Methyl-1-phenylbutan-1-ol ≥96%
3-Methyl-1-phenylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₆O
MW: 164.2441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08442704
CAS Number: 1565-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR