1-(6-Bromo-5-methylpyridin-3-yl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

170059-1G 170059-500MG 170059-5G 170059-25G
77465-794EA 415.09 USD
77465-794 77478-548 77488-746 77499-260
1-(6-Bromo-5-methylpyridin-3-yl)-4,4-dimethylcyclohexanol ≥97%
1-(6-Bromo-5-methylpyridin-3-yl)-4,4-dimethylcyclohexanol

Precaución: For research use only

Formula: C₁₄H₂₀BrNO
MW: 298.2187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34853030
CAS Number: 2921837-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR