1-(6-Bromo-5-methylpyridin-3-yl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

170055-1G 170055-500MG 170055-5G 170055-25G
77465-790EA 415.09 USD
77465-790 77478-544 77488-742 77499-256
1-(6-Bromo-5-methylpyridin-3-yl)cyclobutanol ≥97%
1-(6-Bromo-5-methylpyridin-3-yl)cyclobutanol

Precaución: For research use only

Formula: C₁₀H₁₂BrNO
MW: 242.1123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34853028
CAS Number: 2921869-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR