1-Methoxy-3-[(1-methylethyl)thio]benzene ≥96%

Supplier: AOBChem USA

Warning

170049-1G 170049-500MG 170049-5G
77465-786EA 458.35 USD
77465-786 77478-540 77488-738
1-Methoxy-3-[(1-methylethyl)thio]benzene ≥96%
1-Methoxy-3-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.2825 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621806
CAS Number: 38474-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR