2-Bromo-4-fluoro-1-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

16999-1G 16999-500MG 16999-5G 16999-25G
77465-782EA 327.54 USD
77465-782 77478-536 77488-734 77499-252
2-Bromo-4-fluoro-1-(methoxymethoxy)benzene ≥97%
2-Bromo-4-fluoro-1-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21101704
CAS Number: 474711-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR