6-(cyclopentyloxy)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

169948-1G 169948-500MG 169948-5G 169948-25G
77465-778EA 161.71 USD
77465-778 77478-532 77488-730 77499-248
6-(cyclopentyloxy)nicotinic acid ≥97%
6-(cyclopentyloxy)nicotinic acid

Precaución: For research use only

Formula: C₁₁H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD09740844
CAS Number: 509101-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR