2-(4-Bromo-3-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

16975-1G 16975-5G 16975-25G
77465-770EA 64.89 USD
77465-770 77488-722 77499-240
2-(4-Bromo-3-methoxyphenyl)-1,3-dioxolane ≥97%
2-(4-Bromo-3-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755706
CAS Number: 1807913-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR