1-(5-Bromobenzo[b]thien-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

169597-1G 169597-500MG 169597-5G 169597-25G
77465-766EA 375.95 USD
77465-766 77478-522 77488-718 77499-236
1-(5-Bromobenzo[b]thien-2-yl)ethanone ≥95%
1-(5-Bromobenzo[b]thien-2-yl)ethanone

Precaución: For research use only

Formula: C₁₀H₇BrOS
MW: 255.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28660126
CAS Number: 1423-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR