5-Chloro-2-(propan-2-ylsulfanyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-[(1-methylethyl)thio]pyridine

169374-1G 169374-500MG 169374-5G 169374-25G
77465-762EA 328.57 USD
77465-762 77478-518 77488-710 77499-228
5-Chloro-2-(propan-2-ylsulfanyl)pyridine ≥97%
5-Chloro-2-(propan-2-ylsulfanyl)pyridine

Precaución: For research use only

Formula: C₈H₁₀ClNS
MW: 187.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21641936
CAS Number: 1489477-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR