1-(5-Bromo-4-methyl-2-thienyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

168899-1G 168899-500MG 168899-5G
77465-758EA 497.49 USD
77465-758 77478-514 77488-706
1-(5-Bromo-4-methyl-2-thienyl)-2,2,2-trifluoroethanone ≥95%
1-(5-Bromo-4-methyl-2-thienyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₄BrF₃OS
MW: 273.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107315
CAS Number: 1506267-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR